Assay Details
Target Gene Details
Entrez Gene ID: | 100505386 |
Gene Name: | predicted gene, 21949 |
Gene Aliases: |
Iqcj-schip1, Schip-1-iqcj, Schip1 |
Location: |
Chr.3:67892220-68626482 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm21949 | NM_001113419.2 | NP_001106890.1 | ||
| KJ941154.1 |
Target Gene Details
Entrez Gene ID: | 208426 |
Gene Name: | IQ motif containing J |
Gene Aliases: |
D230050A05, EG208426 |
Location: |
Chr.3:67892220-68056593 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Iqcj | NM_177585.3 | NP_808253.1 | ||
| AK052136.1 | BAC34854.1 | |||
| BC107305.1 | AAI07306.1 | |||
| BC107306.1 | AAI07307.1 |

Genomic Map