Assay Details
Target Gene Details
Entrez Gene ID: | 70099 |
Gene Name: | structural maintenance of chromosomes 4 |
Gene Aliases: |
2500002A22Rik, C79747, SMC-4, Smc4l1 |
Location: |
Chr.3:69004972-69034623 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Smc4 | NM_133786.3 | NP_598547.1 | ||
AJ534940.2 | CAD59183.1 | |||
AK010848.1 | ||||
AK088350.1 | BAC40297.1 | |||
AK088846.1 | BAC40608.2 | |||
AK133817.1 | BAE21860.1 | |||
AK152804.1 | BAE31509.1 | |||
BC061481.1 | AAH61481.1 | |||
BC062939.1 | AAH62939.1 |
