Assay Details
Target Gene Details
Entrez Gene ID: | 329650 |
Gene Name: | mediator complex subunit 12-like |
Gene Aliases: |
A130035F20, A630079M02, AU044581, TRALPUSH |
Location: |
Chr.3:59005595-59318416 on Build GRCm38 |
Assay Gene Location: | Within Intron 34 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Med12l | NM_177855.3 | NP_808523.2 | ||
| AK037666.1 | ||||
| CK638338.1 |
Target Gene Details
Entrez Gene ID: | 70839 |
Gene Name: | purinergic receptor P2Y, G-protein coupled 12 |
Gene Aliases: |
2900079B22Rik, 4921504D23Rik, P2Y12 |
Location: |
Chr.3:59216271-59263005 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| P2ry12 | NM_027571.3 | NP_081847.3 | ||
| AK013804.1 | BAB29000.1 |
More Information
Set Membership: |
|

Genomic Map