Assay Details
Target Gene Details
Entrez Gene ID: | 241989 |
Gene Name: | poly(A) binding protein, cytoplasmic 4-like |
Gene Aliases: |
C330050A14Rik, EG241989 |
Location: |
Chr.3:46442197-46447996 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pabpc4l | NM_001101479.1 | NP_001094949.1 |
