Assay Details
Target Gene Details
Entrez Gene ID: | 59020 |
Gene Name: | PDZ domain containing 1 |
Gene Aliases: |
1700023D20Rik, 2610507N21Rik, 4921513F16Rik, AI267131, AI314638, AL022680, D3Ertd537e, Pdzd1, mPDZK1 |
Location: |
Chr.3:96830065-96870926 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdzk1 | NM_021517.2 | NP_067492.2 | ||
| AF220100.1 | AAF73863.1 | |||
| AK006269.1 | 1 | 465 | BAB24493.1 | |
| AK012070.1 | BAB28007.1 | |||
| AK014879.1 | BAB29600.1 | |||
| AK029752.1 | 1 | 449 | BAC26598.1 | |
| AK029764.1 | 1 | 459 | BAC26605.1 | |
| AK165415.1 | ||||
| AW259368.1 |
More Information
Set Membership: |
|

Genomic Map