Assay Details
Target Gene Details
Entrez Gene ID: | 74414 |
Gene Name: | polymerase (RNA) III (DNA directed) polypeptide C |
Gene Aliases: |
4933407E01Rik, RPC3, RPC62 |
Location: |
Chr.3:96711520-96727480 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Polr3c | NM_028925.1 | NP_083201.1 | ||
| AK016716.1 | BAB30395.1 | |||
| BC147485.1 | ||||
| BC151028.1 |
More Information
Set Membership: |
|

Genomic Map