Assay Details
Target Gene Details
Entrez Gene ID: | 100505386 |
Gene Name: | predicted gene, 21949 |
Gene Aliases: |
Iqcj-schip1, Schip-1-iqcj, Schip1 |
Location: |
Chr.3:67892220-68626482 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm21949 | NM_001113419.2 | 8 | 1444 | NP_001106890.1 |
| KJ941154.1 | 7 | 748 |
Target Gene Details
Entrez Gene ID: | 30953 |
Gene Name: | schwannomin interacting protein 1 |
Gene Aliases: |
Nf2ip, Schip-1 |
Location: |
Chr.3:68064802-68626482 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Schip1 | NM_001113420.1 | 4 | 582 | NP_001106891.1 |
| NM_001113421.1 | 5 | 1266 | NP_001106892.1 | |
| NM_001282044.1 | 5 | 1249 | NP_001268973.1 | |
| NM_001282045.1 | 5 | 1107 | NP_001268974.1 | |
| NM_013928.5 | 4 | 672 | NP_038956.2 | |
| AF145716.1 | 4 | 778 | AAF34244.1 | |
| AK021361.1 | 4 | 533 | BAB32387.1 | |
| AK083464.1 | 5 | 1092 | ||
| AK146930.1 | 5 | 1223 | BAE27541.1 | |
| AK168005.1 | 5 | 1176 | BAE39993.1 | |
| BC060529.1 | 4 | 606 | AAH60529.1 | |
| BC094348.1 | 5 | 1175 | AAH94348.1 | |
| EU163407.1 | 5 | 1266 | ||
| EU163408.1 | 4 | 582 | ||
| KM233715.1 | 4 | 528 | ||
| KM233716.1 | 4 | 533 |

Genomic Map