Assay Details
Target Gene Details
Entrez Gene ID: | 83398 |
Gene Name: | N-deacetylase/N-sulfotransferase (heparan glucosaminyl) 3 |
Gene Aliases: |
4921531K01Rik, 4930511P15Rik, N-HSST 3, NDST-3 |
Location: |
Chr.3:123526166-123690852 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndst3 | NM_001293682.1 | NP_001280611.1 | ||
| NM_031186.3 | NP_112463.2 | |||
| NR_121616.1 | ||||
| AF221095.1 | AAG34793.1 | |||
| AK014990.1 | ||||
| AK015768.1 | BAB29967.1 | |||
| AK141945.1 | BAE24894.1 | |||
| BC079622.1 | AAH79622.1 | |||
| BC112404.1 |
More Information
Set Membership: |
|

Genomic Map