Assay Details
Target Gene Details
Entrez Gene ID: | 18176 |
Gene Name: | neuroblastoma ras oncogene |
Gene Aliases: |
AV095280, N-ras |
Location: |
Chr.3:103058285-103067914 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nras | NM_010937.2 | NP_035067.2 | ||
| AK010412.1 | ||||
| AK011704.1 | BAB27790.1 | |||
| AK150357.1 | BAE29492.1 | |||
| AK165960.1 | BAE38488.1 | |||
| BC051443.1 | AAH51443.1 | |||
| BC058755.1 | AAH58755.1 | |||
| BY321840.1 | ||||
| CT010359.1 | CAJ18567.1 | |||
| X13664.1 | CAA31958.1 |
More Information
Set Membership: |
|

Genomic Map