Assay Details
Target Gene Details
Entrez Gene ID: | 83409 |
Gene Name: | late endosomal/lysosomal adaptor, MAPK and MTOR activator 2 |
Gene Aliases: |
2010111E04Rik, AL022628, Mapbpip, P14, Rab25, Robld3 |
Location: |
Chr.3:88549819-88552927 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lamtor2 | NM_031248.3 | 4 | 486 | NP_112538.1 |
AJ277386.1 | CAB97177.1 | |||
AK019006.1 | 4 | 481 | BAB31506.1 | |
BC031816.1 | 4 | 486 | AAH31816.1 |
