Assay Details
Target Gene Details
Entrez Gene ID: | 20615 |
Gene Name: | SNAP-associated protein |
Gene Aliases: |
25kDa, AA407989, AV026596, Bloc1s7, Snap25bp, Snapap |
Location: |
Chr.3:90488026-90491013 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Snapin | NM_133854.3 | 1 | 50 | NP_598615.1 |
AF086838.1 | AAD11418.1 | |||
AK007458.1 | BAB25049.1 | |||
AK035147.1 | 1 | 52 | ||
AK044085.1 | ||||
AK048007.1 | 1 | 39 | BAC33212.1 | |
AK050151.1 | 1 | 51 | BAC34095.1 | |
AK077840.1 | 1 | 41 | BAC37029.1 | |
AK080718.1 | 1 | 41 | BAC37991.1 | |
AK135006.1 | 1 | 42 | BAE22381.1 | |
AK152057.1 | BAE30913.1 | |||
BC048691.1 | AAH48691.1 |
