Assay Details
Target Gene Details
Entrez Gene ID: | 56426 |
Gene Name: | programmed cell death 10 |
Gene Aliases: |
2410003B13Rik, Ccm3, Tfa15, Tfar15 |
Location: |
Chr.3:75516490-75556852 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pdcd10 | NM_019745.4 | NP_062719.2 | ||
AF159368.1 | AAD44335.1 | |||
AK005594.1 | BAB24143.1 | |||
AK075926.1 | BAC36058.1 | |||
AK077308.1 | BAC36742.1 | |||
AK137198.1 | BAE23263.1 | |||
AK149182.1 | ||||
AK160234.1 | BAE35705.1 | |||
AK168857.1 | BAE40678.1 | |||
AK169689.1 | BAE41306.1 | |||
BC019650.1 | AAH19650.1 | |||
BC086778.1 | AAH86778.1 | |||
BY091056.1 |
