Assay Details
Target Gene Details
Entrez Gene ID: | 22035 |
Gene Name: | tumor necrosis factor (ligand) superfamily, member 10 |
Gene Aliases: |
A330042I21Rik, AI448571, APO-2L, Ly81, TL2, Trail |
Location: |
Chr.3:27317077-27339665 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnfsf10 | NM_009425.2 | 2 | 277 | NP_033451.1 |
| AK142960.1 | 2 | 279 | BAE25231.1 | |
| AK153581.1 | 2 | 327 | BAE32108.1 | |
| BC138807.1 | 2 | 251 | ||
| BC138808.1 | 2 | 251 | ||
| BY734631.1 | 2 | 294 | ||
| U37522.1 | 2 | 253 | AAC52345.1 |

Genomic Map