Assay Details
Target Gene Details
Entrez Gene ID: | 28030 |
Gene Name: | G elongation factor, mitochondrial 1 |
Gene Aliases: |
AW545374, D3Wsu133e, Gfm |
Location: |
Chr.3:67430115-67475068 on Build GRCm38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gfm1 | NM_138591.2 | NP_613057.2 | ||
| AF315511.1 | AAK58878.1 | |||
| BC013093.1 | AAH13093.1 | |||
| BC031772.1 | AAH31772.1 |
Target Gene Details
Entrez Gene ID: | 17035 |
Gene Name: | latexin |
Gene Aliases: |
- |
Location: |
Chr.3:67457999-67463907 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lxn | NM_016753.4 | NP_058033.2 | ||
| AK032170.1 | BAC27739.1 | |||
| AK149981.1 | BAE29211.1 | |||
| BC115521.1 | ||||
| BC115522.1 | ||||
| D88769.1 | BAA13700.1 |
More Information
Set Membership: |
|

Genomic Map