Assay Details
Target Gene Details
Entrez Gene ID: | 19055 |
Gene Name: | protein phosphatase 3, catalytic subunit, alpha isoform |
Gene Aliases: |
2900074D19Rik, CN, Caln, Calna, CnA |
Location: |
Chr.3:136670066-136937727 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp3ca | NM_001293622.1 | 2 | 896 | NP_001280551.1 |
| NM_008913.5 | 2 | 896 | NP_032939.1 | |
| AK146387.1 | 2 | 840 | BAE27131.1 | |
| AK150393.1 | 2 | 255 | BAE29521.1 | |
| AK166287.1 | 2 | 671 | ||
| AK209023.1 | 1 | 37 | ||
| BC096652.1 | 2 | 138 | AAH96652.1 | |
| BC138612.1 | 2 | 169 | ||
| J05479.1 | 2 | 193 | AAA37359.1 |
More Information
Set Membership: |
|

Genomic Map