Assay Details
Target Gene Details
Entrez Gene ID: | 19055 |
Gene Name: | protein phosphatase 3, catalytic subunit, alpha isoform |
Gene Aliases: |
2900074D19Rik, CN, Caln, Calna, CnA |
Location: |
Chr.3:136670066-136937727 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp3ca | NM_001293622.1 | NP_001280551.1 | ||
| NM_008913.5 | NP_032939.1 | |||
| AK146387.1 | BAE27131.1 | |||
| AK150393.1 | BAE29521.1 | |||
| AK166287.1 | ||||
| AK177425.1 | ||||
| AK177666.1 | ||||
| AK189167.1 | ||||
| AK191293.1 | ||||
| AK191345.1 | ||||
| AK197466.1 | ||||
| AK206754.1 | ||||
| AK211949.1 | ||||
| AK211995.1 | ||||
| AK213582.1 | ||||
| AK219430.1 | ||||
| BC096652.1 | AAH96652.1 | |||
| BC138612.1 | ||||
| J04134.1 | AAA37432.1 | |||
| J05479.1 | AAA37359.1 |
More Information
Set Membership: |
|

Genomic Map