Assay Details
Target Gene Details
Entrez Gene ID: | 83679 |
Gene Name: | phosphodiesterase 4D interacting protein (myomegalin) |
Gene Aliases: |
4732458A06Rik, 9430063L05Rik, C87016, D130016K21Rik, D3Bwg1078e, Usmg4, mKIAA0454 |
Location: |
Chr.3:97689828-97888707 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 42 - Exon 42 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pde4dip | NM_001039376.2 | NP_001034465.2 | ||
NM_001289701.1 | NP_001276630.1 | |||
NM_001289702.1 | NP_001276631.1 | |||
AK040750.1 | BAC30693.1 | |||
BB636676.1 | ||||
BC141185.1 | ||||
BC145504.1 | ||||
BM950385.1 |
