Assay Details
Target Gene Details
Entrez Gene ID: | 83679 |
Gene Name: | phosphodiesterase 4D interacting protein (myomegalin) |
Gene Aliases: |
4732458A06Rik, 9430063L05Rik, C87016, D130016K21Rik, D3Bwg1078e, Usmg4, mKIAA0454 |
Location: |
Chr.3:97689828-97888707 on Build GRCm38 |
Assay Gene Location: | Within Exon 44 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pde4dip | NM_001039376.2 | 43 | 6902 | NP_001034465.2 |
NM_001289701.1 | 43 | 6953 | NP_001276630.1 | |
NM_001289702.1 | 42 | 6800 | NP_001276631.1 | |
AK040750.1 | 4 | 540 | BAC30693.1 | |
BC141185.1 | 43 | 6953 | ||
BC145504.1 | 42 | 6800 | ||
BM950385.1 | 3 | 458 |
