Assay Details
Target Gene Details
Entrez Gene ID: | 230393 |
Gene Name: | focadhesin |
Gene Aliases: |
Kiaa1797, mKIAA1797 |
Location: |
Chr.4:88094630-88411011 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Focad | NM_001081184.1 | NP_001074653.1 | ||
| BC057079.1 | AAH57079.1 |

Genomic Map