Assay Details
Target Gene Details
Entrez Gene ID: | 73398 |
Gene Name: | nipsnap homolog 3A (C. elegans) |
Gene Aliases: |
1700054F22Rik, AI429120 |
Location: |
Chr.4:52989284-53000854 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nipsnap3a | NM_028529.1 | NP_082805.1 | ||
| AK006782.1 | BAB24737.1 | |||
| AV260630.1 | ||||
| BY465731.1 |

Genomic Map