Assay Details
Target Gene Details
Entrez Gene ID: | 66105 |
Gene Name: | ubiquitin-conjugating enzyme E2D 3 |
Gene Aliases: |
1100001F19Rik, 9430029A22Rik, AA414951 |
Location: |
Chr.3:135438244-135467180 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ube2d3 | NM_025356.4 | NP_079632.1 | ||
AK003163.1 | BAB22614.1 | |||
AK004001.1 | BAB23116.1 | |||
AK009276.1 | BAB26188.1 | |||
AK009936.1 | ||||
AK014344.1 | ||||
AK032885.1 | BAC28070.1 | |||
AK049906.1 | BAC33981.1 | |||
AK077666.1 | BAC36940.1 | |||
AK088448.1 | BAC40357.1 | |||
AK151295.1 | BAE30279.1 | |||
AK151594.1 | BAE30534.1 | |||
AK152231.1 | BAE31058.1 | |||
AK152541.1 | BAE31296.1 | |||
AK153100.1 | BAE31721.1 | |||
AK153111.1 | BAE31728.1 | |||
AK153410.1 | BAE31970.1 | |||
AK153423.1 | ||||
AK159125.1 | BAE34839.1 | |||
AK159546.1 | BAE35172.1 | |||
AK159963.1 | BAE35518.1 | |||
AK165235.1 | BAE38093.1 | |||
AK167077.1 | BAE39237.1 | |||
AK167138.1 | BAE39283.1 | |||
AK167650.1 | BAE39701.1 | |||
AK167801.1 | BAE39829.1 | |||
AK169177.1 | BAE40955.1 | |||
BC024351.1 | ||||
BC057941.1 | AAH57941.1 | |||
BC098220.1 | AAH98220.1 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
