Assay Details
Target Gene Details
Entrez Gene ID: | 66291 |
Gene Name: | small integral membrane protein 8 |
Gene Aliases: |
1810030N24Rik, 2810406B13Rik |
Location: |
Chr.4:34768664-34778396 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Smim8 | NM_025471.2 | NP_079747.1 | ||
AK005941.1 | BAB24328.1 | |||
AK007303.1 | BAB24944.1 | |||
AK007660.1 | BAB25170.1 | |||
AK013007.1 | 1 | 108 | ||
AK021236.1 | BAB32341.1 | |||
AK079613.1 | ||||
BC027508.1 | AAH27508.1 |
