Assay Details
Target Gene Details
| Entrez Gene ID: | 654812 | 
| Gene Name: | angiopoietin-like 7 | 
| Gene Aliases: | - | 
| Location: | Chr.4:148492979-148500462 on Build GRCm38 | 
| Assay Gene Location: | Within Exon 3 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| Angptl7 | NM_001039554.3 | 3 | 728 | NP_001034643.1 | 
| AK031675.1 | 3 | 728 | BAC27507.1 | |
| BC023373.1 | 3 | 715 | AAH23373.1 | 
Target Gene Details
| Entrez Gene ID: | 56717 | 
| Gene Name: | mechanistic target of rapamycin (serine/threonine kinase) | 
| Gene Aliases: | 2610315D21Rik, AI327068, FRAP, FRAP2, Frap1, RAFT1, RAPT1, flat | 
| Location: | Chr.4:148448582-148557685 on Build GRCm38 | 
| Assay Gene Location: | Within Intron 28 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| Mtor | NM_020009.2 | NP_064393.2 | ||
| AF152838.1 | AAF73196.1 | |||
| AK143381.1 | ||||
| AK158715.1 | ||||
| BC112904.1 | AAI12905.1 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Exonic  Intronic  Non-exonic | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top