Assay Details
Target Gene Details
Entrez Gene ID: | 329828 |
Gene Name: | expressed sequence AI464131 |
Gene Aliases: |
AA408153, AI604836, Gm762, Kiaa1161, NET37 |
Location: |
Chr.4:41491847-41503136 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AI464131 | NM_001085515.2 | 1 | 41 | NP_001078984.1 |
| AK173117.1 | BAD32395.1 | |||
| BC137640.1 | ||||
| BC140956.1 | ||||
| BY246173.1 | ||||
| BY283520.1 | 1 | 41 |
More Information
Set Membership: |
|

Genomic Map