Assay Details
Target Gene Details
Entrez Gene ID: | 329954 |
Gene Name: | cation channel, sperm associated 4 |
Gene Aliases: |
4933412L05 |
Location: |
Chr.4:134211968-134227434 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Catsper4 | NM_001130030.1 | 8 | 1030 | NP_001123502.1 |
NM_177866.4 | 10 | 1249 | NP_808534.1 | |
AK077145.1 | 10 | 1236 | BAC36641.1 | |
BC108974.2 | 10 | 1229 | AAI08975.1 | |
BC108975.1 | 10 | 1229 | AAI08976.1 | |
EU781510.1 | 9 | 1126 | ||
EU781511.1 | 8 | 1005 |
