Assay Details
Target Gene Details
Entrez Gene ID: | 12557 |
Gene Name: | cadherin 17 |
Gene Aliases: |
HPT-1, HPT-1/LI |
Location: |
Chr.4:11758157-11817905 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdh17 | NM_019753.4 | 8 | 992 | NP_062727.1 |
| AF177669.1 | 8 | 888 | AAD51125.1 | |
| AK089791.1 | 8 | 959 | ||
| AK155633.1 | 8 | 993 | BAE33356.1 | |
| AK171733.1 | 8 | 915 | BAE42638.1 | |
| BC042891.1 | 8 | 910 | AAH42891.1 | |
| D87912.1 | 8 | 937 | BAB03264.1 |
More Information
Set Membership: |
|

Genomic Map