Assay Details
Target Gene Details
Entrez Gene ID: | 74183 |
Gene Name: | PPARGC1 and ESRR induced regulator, muscle 1 |
Gene Aliases: |
2310042D19Rik |
Location: |
Chr.4:156203268-156221307 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Perm1 | NM_172417.3 | NP_766005.2 | ||
| AK036660.1 | BAC29522.1 | |||
| AK040833.1 | BAC30714.1 |

Genomic Map