Assay Details
Target Gene Details
Entrez Gene ID: | 66665 |
Gene Name: | Myb/SANT-like DNA-binding domain containing 3 |
Gene Aliases: |
2410046L14Rik, 2810429P03Rik, 5730528L13Rik, Mysantd3 |
Location: |
Chr.4:48539832-48565892 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Msantd3 | NM_001145924.1 | NP_001139396.1 | ||
| NM_001145925.1 | NP_001139397.1 | |||
| AK011381.1 | BAB27581.1 | |||
| AK017795.1 | BAB30936.1 | |||
| AK075956.1 | BAC36081.1 |

Genomic Map