Assay Details
Target Gene Details
Entrez Gene ID: | 67770 |
Gene Name: | caspase activity and apoptosis inhibitor 1 |
Gene Aliases: |
5830433M19Rik, AI536248, CAAP |
Location: |
Chr.4:94460277-94557076 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Caap1 | NM_026368.2 | NP_080644.2 | ||
AK076487.1 | ||||
AK140382.1 | BAE24364.1 | |||
AK150311.1 | ||||
AK160297.1 | ||||
AK160572.1 | BAE35881.1 | |||
AK171141.1 | ||||
BC020067.1 | AAH20067.1 |
