Assay Details
Target Gene Details
Entrez Gene ID: | 77032 |
Gene Name: | thiosulfate sulfurtransferase (rhodanese)-like domain containing 3 |
Gene Aliases: |
2610029I01Rik |
Location: |
Chr.4:21757382-21767211 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tstd3 | NM_029840.1 | 1 | 42 | NP_084116.1 |
| AI287097.1 | 1 | 42 | ||
| AK011611.1 | BAB27732.1 | |||
| AK154853.1 | BAE32878.1 |
Target Gene Details
Entrez Gene ID: | 77593 |
Gene Name: | ubiquitin specific petidase 45 |
Gene Aliases: |
3110003C05Rik, 4930550B20Rik, AI843191, Gcap7 |
Location: |
Chr.4:21767161-21837872 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Usp45 | AK019756.1 |
More Information
Set Membership: |
|

Genomic Map