Assay Details
Target Gene Details
Entrez Gene ID: | 51813 |
Gene Name: | cyclin C |
Gene Aliases: |
AI451004, AU020987, CG1C |
Location: |
Chr.4:21727701-21759922 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccnc | NM_001290420.1 | 12 | 1216 | NP_001277349.1 |
| AK135472.1 | 12 | 1212 | BAE22544.1 |
Target Gene Details
Entrez Gene ID: | 77032 |
Gene Name: | thiosulfate sulfurtransferase (rhodanese)-like domain containing 3 |
Gene Aliases: |
2610029I01Rik |
Location: |
Chr.4:21757382-21767211 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tstd3 | NM_029840.1 | NP_084116.1 | ||
| AI287097.1 | ||||
| AK011611.1 | BAB27732.1 | |||
| AK154853.1 | BAE32878.1 | |||
| AK218953.1 |
More Information
Set Membership: |
|

Genomic Map