Assay Details
Target Gene Details
Entrez Gene ID: | 76166 |
Gene Name: | REM2 and RAB-like small GTPase 1 |
Gene Aliases: |
6330545A04Rik, Gm723 |
Location: |
Chr.4:141213666-141220365 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rsg1 | NM_001081174.2 | NP_001074643.1 | ||
| BY232166.1 |

Genomic Map