Assay Details
Target Gene Details
Entrez Gene ID: | 77739 |
Gene Name: | ADAMTS-like 1 |
Gene Aliases: |
5930437A14Rik, 6720426B09Rik, ADAMTSL-1 |
Location: |
Chr.4:85513234-86428390 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adamtsl1 | NM_029967.3 | 21 | 4084 | NP_084243.3 |
| AK031246.1 | 4 | 687 | BAC27317.1 | |
| AK132420.1 | 9 | 2524 | BAE21159.1 | |
| BC116295.1 | 4 | 678 | ||
| BC116296.1 | 4 | 678 |
More Information
Set Membership: |
|

Genomic Map