Assay Details
Target Gene Details
Entrez Gene ID: | 11641 |
Gene Name: | A kinase (PRKA) anchor protein 2 |
Gene Aliases: |
AA959716, AI649048, Akap-2, Akap-kl, B230340M18Rik, Prka2 |
Location: |
Chr.4:57845248-57896984 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Akap2 | NM_001035532.2 | NP_001030609.1 | ||
NM_001035533.2 | NP_001030610.1 | |||
NM_009649.3 | NP_033779.2 | |||
AF033274.1 | AAC02206.1 | |||
AF033275.1 | AAC02207.1 | |||
AF033276.1 | AAC02208.1 | |||
AK041873.1 | BAC31088.1 | |||
AK147420.1 | BAE27901.1 | |||
AK162156.1 | ||||
AK202562.1 | ||||
AK215055.1 | ||||
AW489512.1 | ||||
BC003735.1 | AAH03735.1 |
