Assay Details
Target Gene Details
Entrez Gene ID: | 12017 |
Gene Name: | BCL2-associated athanogene 1 |
Gene Aliases: |
BAG-1, Rap46 |
Location: |
Chr.4:40936398-40948294 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bag1 | NM_001171739.1 | 2 | 645 | NP_001165210.1 |
NM_009736.3 | 2 | 645 | NP_033866.4 | |
AF022223.1 | 2 | 586 | AAC34259.1 | |
AK009149.1 | 2 | 556 | BAB26106.1 | |
AK154469.1 | 2 | 595 | ||
AK171094.1 | 2 | 595 | ||
BC003722.1 | 2 | 607 | ||
BC069918.1 | 2 | 582 | ||
BC093509.1 | 2 | 556 | ||
CA321856.1 | 2 | 632 | ||
CX225744.1 | 2 | 645 |
