Assay Details
Target Gene Details
Entrez Gene ID: | 78784 |
Gene Name: | CUGBP, Elav-like family member 3 |
Gene Aliases: |
4930415M08Rik, BRUNOL1, CAGH4, ERDA4, Tnrc4 |
Location: |
Chr.3:94478285-94492199 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Celf3 | NM_001289613.1 | NP_001276542.1 | ||
| NM_001289616.1 | NP_001276545.1 | |||
| NM_001289620.1 | NP_001276549.1 | |||
| NM_172434.3 | NP_766022.1 | |||
| AK044069.1 | ||||
| AK044506.1 | ||||
| AK087536.1 | ||||
| AY165052.1 | AAN73885.1 | |||
| BC057553.1 | AAH57553.1 | |||
| BY089288.1 |

Genomic Map