Assay Details
Target Gene Details
Entrez Gene ID: | 78266 |
Gene Name: | zinc finger protein 687 |
Gene Aliases: |
4931408L03Rik, Znf687, mKIAA1441 |
Location: |
Chr.3:95006702-95015452 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp687 | NM_030074.2 | 4 | 2574 | NP_084350.1 |
| AK019851.2 | 4 | 2575 | BAB31881.1 | |
| AK129360.1 | 2 | 2301 | BAC98170.1 | |
| AK133173.1 | 3 | 3085 | ||
| AK148239.1 | 4 | 2531 | ||
| AK207582.1 | 1 | 158 | ||
| BC056943.1 | AAH56943.1 | |||
| BC060179.1 | AAH60179.1 |

Genomic Map