Assay Details
Target Gene Details
Entrez Gene ID: | 78266 |
Gene Name: | zinc finger protein 687 |
Gene Aliases: |
4931408L03Rik, Znf687, mKIAA1441 |
Location: |
Chr.3:95006702-95015452 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp687 | NM_030074.2 | NP_084350.1 | ||
| AK019851.2 | BAB31881.1 | |||
| AK129360.1 | 3 | 3415 | BAC98170.1 | |
| AK133173.1 | 6 | 3938 | ||
| AK148239.1 | ||||
| BC056943.1 | AAH56943.1 | |||
| BC060179.1 | AAH60179.1 |

Genomic Map