Assay Details
Target Gene Details
Entrez Gene ID: | 381572 |
Gene Name: | RIKEN cDNA 9430007A20 gene |
Gene Aliases: |
Aadacl4, Gm1034 |
Location: |
Chr.4:144519822-144529353 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 9430007A20Rik | NM_198662.3 | NP_941064.2 | ||
| AK034567.1 | BAC28757.1 | |||
| BC148288.1 |
More Information
Set Membership: |
|

Genomic Map