Assay Details
Target Gene Details
Entrez Gene ID: | 58810 |
Gene Name: | aldo-keto reductase family 1, member A1 (aldehyde reductase) |
Gene Aliases: |
2610201A18Rik, Akr1a4 |
Location: |
Chr.4:116636510-116651674 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Akr1a1 | NM_021473.3 | NP_067448.1 | ||
| AF225564.1 | AAF67111.1 | |||
| AK005376.1 | ||||
| AK009462.1 | BAB26303.1 | |||
| AK011321.1 | BAB27543.1 | |||
| AK011388.1 | BAB27586.1 | |||
| AK011667.1 | BAB27767.1 | |||
| AK011794.1 | BAB27846.1 | |||
| AK011856.1 | BAB27883.1 | |||
| AK011906.1 | BAB27907.1 | |||
| AK011908.1 | BAB27909.1 | |||
| AK011918.1 | BAB27915.1 | |||
| AK128906.1 | ||||
| AK144198.1 | BAE25762.1 | |||
| AK146081.1 | BAE26885.1 | |||
| AK146278.1 | BAE27036.1 | |||
| AK152086.1 | BAE30936.1 | |||
| AK159573.1 | ||||
| AK160185.1 | ||||
| AK164461.1 | BAE37798.1 | |||
| AK168950.1 | BAE40756.1 | |||
| AK186333.1 | ||||
| AK192030.1 | ||||
| AK210646.1 | ||||
| AK219186.1 | ||||
| AY207463.1 | AAO72145.1 | |||
| BC039926.1 | AAH39926.1 | |||
| BC046762.1 | AAH46762.1 |
More Information
Set Membership: |
|

Genomic Map