Assay Details
Target Gene Details
Entrez Gene ID: | 53861 |
Gene Name: | zinc finger, RAN-binding domain containing 2 |
Gene Aliases: |
AI227013, Zfp265, Zis, Znf265 |
Location: |
Chr.3:157534397-157548339 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zranb2 | NM_017381.2 | NP_059077.1 | ||
| AF133818.1 | AAF04474.1 | |||
| AK146275.1 | BAE27034.1 | |||
| AK146854.1 | ||||
| AK150291.1 | BAE29446.1 | |||
| AK152536.1 | BAE31292.1 | |||
| AK160862.1 | BAE36054.1 | |||
| AK166534.1 | BAE38837.1 | |||
| AK167756.1 | BAE39790.1 | |||
| AK167766.1 | BAE39799.1 | |||
| AK167862.1 | BAE39880.1 | |||
| AK167983.1 | BAE39974.1 | |||
| AK168033.1 | BAE40017.1 | |||
| AK168162.1 | BAE40123.1 | |||
| AK168613.1 | BAE40479.1 | |||
| AK169114.1 | BAE40895.1 | |||
| AK169335.1 | BAE41087.1 | |||
| BC051187.1 | ||||
| BC132547.1 | ||||
| BC138575.1 |
More Information
Set Membership: |
|

Genomic Map