Assay Details
Target Gene Details
Entrez Gene ID: | 14612 |
Gene Name: | gap junction protein, alpha 4 |
Gene Aliases: |
AU020209, AW558810, Cnx37, Cx37, Gja-4 |
Location: |
Chr.4:127311420-127314039 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gja4 | NM_008120.3 | NP_032146.1 | ||
| AF216832.1 | AAF91221.1 | |||
| AK143024.1 | BAE25252.1 | |||
| BC056613.1 | AAH56613.1 |
More Information
Set Membership: |
|

Genomic Map