Assay Details
Target Gene Details
Entrez Gene ID: | 14620 |
Gene Name: | gap junction protein, beta 3 |
Gene Aliases: |
Cnx31, Cx31, D4Wsu144e, Gjb-3 |
Location: |
Chr.4:127325235-127330836 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gjb3 | NM_001160012.1 | NP_001153484.1 | ||
| NM_008126.2 | NP_032152.1 | |||
| AK083716.1 | BAC39002.1 | |||
| AY390400.1 | AAQ93033.1 | |||
| BC024387.1 | AAH24387.1 | |||
| BY027476.1 | ||||
| CJ084102.1 | ||||
| X63099.1 | CAA44813.1 |
More Information
Set Membership: |
|

Genomic Map