Assay Details
Target Gene Details
Entrez Gene ID: | 14621 |
Gene Name: | gap junction protein, beta 4 |
Gene Aliases: |
Cnx30.3, Cx30.3, Gjb-4 |
Location: |
Chr.4:127331418-127354141 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gjb4 |
