Assay Details
Target Gene Details
Entrez Gene ID: | 667250 |
Gene Name: | predicted gene 12657 |
Gene Aliases: |
OTTMUSG00000007855 |
Location: |
Chr.4:94593650-94600319 on Build GRCm38 |
Assay Gene Location: | Overlaps - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm12657 | NM_001081019.1 | NP_001074488.1 |
Target Gene Details
Entrez Gene ID: | 18786 |
Gene Name: | phospholipase A2, activating protein |
Gene Aliases: |
2410007N06, AI536418, AU018445, AW208417, D4Ertd618e, PLA2P, PLAP, Ufd3 |
Location: |
Chr.4:94565139-94603247 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Plaa | NM_172695.2 | NP_766283.2 | ||
| AK019070.1 | ||||
| AK075934.1 | BAC36064.1 | |||
| AK081641.1 | ||||
| AK162534.1 | ||||
| AK166122.1 | ||||
| BC139355.1 | ||||
| BC139356.1 | ||||
| BC139773.1 | ||||
| CA490654.1 |
More Information
Set Membership: |
|

Genomic Map