Assay Details
Target Gene Details
Entrez Gene ID: | 13806 |
Gene Name: | enolase 1, alpha non-neuron |
Gene Aliases: |
0610008I15, AL022784, Eno-1, MBP-1 |
Location: |
Chr.4:150236726-150248873 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eno1 | NM_023119.2 | NP_075608.2 | ||
| AK002322.1 | BAC24987.1 | |||
| AK002336.1 | BAB22021.1 | |||
| AK088788.1 | BAC40572.1 | |||
| AK147987.1 | BAE28270.1 | |||
| AK150161.1 | BAE29351.1 | |||
| AK150192.1 | BAE29369.1 | |||
| AK153321.1 | BAE31901.1 | |||
| AK191162.1 | ||||
| AK191332.1 | ||||
| AK196981.1 | ||||
| AK198034.1 | ||||
| AK199229.1 | ||||
| AK202164.1 | ||||
| AK202428.1 | ||||
| AK202509.1 | ||||
| AK203149.1 | ||||
| AK204087.1 | ||||
| AK204457.1 | ||||
| AK204949.1 | ||||
| AK214077.1 | ||||
| BC003891.1 | AAH03891.1 | |||
| BC010685.1 | AAH10685.1 | |||
| BC024644.1 | AAH24644.1 | |||
| BC039179.1 | ||||
| BC056611.1 | AAH56611.1 | |||
| BC083334.1 | ||||
| BC085098.1 | AAH85098.1 | |||
| BU526382.1 | ||||
| X52379.1 | CAA36605.1 |
More Information
Set Membership: |
|

Genomic Map