Assay Details
Target Gene Details
Entrez Gene ID: | 57756 |
Gene Name: | four and a half LIM domains 5 |
Gene Aliases: |
1700027G07Rik, Act |
Location: |
Chr.4:25199907-25243044 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fhl5 | NM_021318.3 | NP_067293.1 | ||
| AF083394.1 | AAD23916.1 | |||
| AK006414.1 | BAB24576.1 | |||
| BC048473.1 | AAH48473.1 |
More Information
Set Membership: |
|

Genomic Map