Assay Details
Target Gene Details
Entrez Gene ID: | 230579 |
Gene Name: | family with sequence simliarity 151, member A |
Gene Aliases: |
2010309H15Rik |
Location: |
Chr.4:106733915-106748292 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fam151a | NM_146149.1 | 1 | 44 | NP_666261.1 |
AY168619.1 | 1 | 44 | AAO38748.1 | |
BC024426.1 | 1 | 44 | AAH24426.1 | |
BC024430.1 | 1 | 44 | AAH24430.1 | |
BC026682.1 | 1 | 53 | AAH26682.1 |
Target Gene Details
Entrez Gene ID: | 381538 |
Gene Name: | maestro heat-like repeat family member 7 |
Gene Aliases: |
Gm1027, Heatr8 |
Location: |
Chr.4:106680417-106736620 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mroh7 |
