Assay Details
Target Gene Details
Entrez Gene ID: | 230579 |
Gene Name: | family with sequence simliarity 151, member A |
Gene Aliases: |
2010309H15Rik |
Location: |
Chr.4:106733915-106748292 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam151a | NM_146149.1 | NP_666261.1 | ||
| AY168619.1 | AAO38748.1 | |||
| BC024426.1 | AAH24426.1 | |||
| BC024430.1 | AAH24430.1 | |||
| BC026682.1 | AAH26682.1 |

Genomic Map