Assay Details
Target Gene Details
Entrez Gene ID: | 76824 |
Gene Name: | mitochondrial fission regulator 1-like |
Gene Aliases: |
2410166I05Rik, AI480750, Fam54b |
Location: |
Chr.4:134525555-134535387 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mtfr1l | NM_001256112.1 | NP_001243041.1 | ||
| NM_029759.4 | NP_084035.1 | |||
| AK010825.1 | BAB27207.1 | |||
| AK080126.1 | ||||
| AK089141.1 | BAC40763.1 | |||
| AK145362.1 | BAE26389.1 | |||
| AK169047.1 | BAE40835.1 | |||
| AK171703.1 | BAE42619.1 | |||
| BY094859.1 | ||||
| BY123213.1 |

Genomic Map